Tisk…

5-Aminonicotinic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Aminopyridine-3-carboxylic acid, 5-Amino-3-pyridinecarboxylic acid

672513-1G 672513-5G
SIAL672513-1GEA 4320 CZK
SIAL672513-1G SIAL672513-5G
5-Aminonicotinic acid, Sigma-Aldrich®
5-Aminonicotinic acid
Vzorec: C₆H₆N₂O₂
MW: 138,13 g/mol
MDL Number: MFCD00129116
CAS číslo: 24242-19-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR