Tisk…

2,6-Dichloro-4-nitropyridine, Sigma-Aldrich®

Dodavatel: Merck

647411-1G 647411-5G
SIAL647411-1GEA 0 CZK
SIAL647411-1G SIAL647411-5G
2,6-Dichloro-4-nitropyridine, Sigma-Aldrich®
2,6-Dichloro-4-nitropyridine
Vzorec: C₅H₂Cl₂N₂O₂
MW: 192,99 g/mol
MDL Number: MFCD05670545
CAS číslo: 25194-01-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR