Tisk…

Methyl-6-chloronicotinate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 6-chloronicotinate

631175-25G 631175-5G
SIAL631175-25GEA 6610 CZK
SIAL631175-25G SIAL631175-5G
Methyl-6-chloronicotinate, Sigma-Aldrich®
Methyl-6-chloronicotinate
Vzorec: C₇H₆ClNO₂
MW: 171,58 g/mol
MDL Number: MFCD00023420
CAS číslo: 73781-91-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR