Tisk…

Diphenyliodonium hexafluorophosphate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

548014-25G 548014-5G
SIAL548014-25GEA 17180 CZK
SIAL548014-25G SIAL548014-5G
Diphenyliodonium hexafluorophosphate, Sigma-Aldrich®
Diphenyliodonium hexafluorophosphate
Vzorec: C₁₂H₁₀F₆IP
MW: 426,08 g/mol
MDL Number: MFCD00061398
CAS číslo: 58109-40-3
EINECS: 261-134-5
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR