(S)-(-)-Epoxystyrene, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(S)-Styrene oxide
M-Clarity™ quality level = MQ100
540102-25G
540102-5G
SIAL540102-25GEA
21860
CZK
SIAL540102-25G
SIAL540102-5G
(S)-(-)-Epoxystyrene, Sigma-Aldrich®
(S)-(-)-Epoxystyrene
Vzorec:
C₈H₈O MW: 120.15 |
MDL Number:
MFCD00064310 CAS číslo: 20780-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...