Tisk…

2-Chloronicotinonitrile, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Chloro-3-cyanopyridine, 2-Chloropyridine-3-carbonitrile

M-Clarity™ quality level = MQ100

535338-25G 535338-500G
SIAL535338-25GEA 1150 CZK
SIAL535338-25G SIAL535338-500G
2-Chloronicotinonitrile, Sigma-Aldrich®
2-Chloronicotinonitrile
Vzorec: C₆H₃ClN₂
MW: 138.56
Bod varu: 112 °C (1 mmHg)
Bod tání: 107…108 °C
MDL Number: MFCD00014628
CAS číslo: 6602-54-6
EINECS: 229-550-1
UN: 3439
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR