Tisk…

3-Methylpicolinic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Methylpyridine-2-carboxylic acid

512362-1G 512362-5G
SIAL512362-1GEA 882 CZK
SIAL512362-1G SIAL512362-5G
3-Methylpicolinic acid, Sigma-Aldrich®
3-Methylpicolinic acid
Vzorec: C₇H₇NO₂
MW: 137,14 g/mol
MDL Number: MFCD00191553
CAS číslo: 4021-07-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR