Tisk…

Sodium methanesulphinate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methanesulphinic acid, sodium salt

M-Clarity™ quality level = MQ100

433063-10G 433063-1G
SIAL433063-10GEA 3120 CZK
SIAL433063-10G SIAL433063-1G
Sodium methanesulphinate, Sigma-Aldrich®
Sodium methanesulphinate
Vzorec: CH₃NaO₂S
MW: 102.09
MDL Number: MFCD00040392
CAS číslo: 20277-69-4
EINECS: 243-669-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR