Tisk…

2-Mercaptonicotinic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Thionicotinic acid

M-Clarity™ quality level = MQ200

419702-100G 419702-25G
SIAL419702-100GEA 4670 CZK
SIAL419702-100G SIAL419702-25G
2-Mercaptonicotinic acid, Sigma-Aldrich®
2-Mercaptonicotinic acid
Vzorec: C₆H₅NO₂S
MDL Number: MFCD00010102
CAS číslo: 38521-46-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR