Polyphosphoric acid trimethylsilyl ester, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
414026-100ML
414026-25ML
SIAL414026-100MLEA
11500
CZK
SIAL414026-100ML
SIAL414026-25ML
Polyphosphoric acid trimethylsilyl ester, Sigma-Aldrich®
Polyphosphoric acid trimethylsilyl ester
Vzorec:
H(PO₂(OSi(CH₃)₃))nOH |
CAS číslo:
40623-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...