Tisk…
Paladium 30% on activated carbon for use as a catalyst, Sigma-Aldrich®
Dodavatel: Merck
Varování
M-Clarity™ quality level = MQ100
407305-10G 407305-1G
SIAL407305-10GEA 20920 CZK
SIAL407305-10G SIAL407305-1G
Paladium 30% on activated carbon for use as a catalyst, Sigma-Aldrich®
Palladium
Vzorec: Pd
MW: 106.420 g/mol
MDL Number: MFCD00011167
CAS číslo: 7440-05-3
EINECS: 231-115-6