Tisk…
Aminoguanidine hydrochloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Aminoguanidine HCl, Aminoguanidine Hydrochloride, Guanylhydrazine hydrochloride
396494-100G 396494-25G
SIAL396494-100GEA 6080 CZK
SIAL396494-100G SIAL396494-25G
Aminoguanidine hydrochloride, Sigma-Aldrich®
Aminoguanidine hydrochloride
Vzorec: CH₇ClN₄
MW: 110,55 g/mol
MDL Number: MFCD00039074
CAS číslo: 1937-19-5
EINECS: 217-707-7