6-Hydroxypicolinic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
6-Hydroxy-2-pyridinecarboxylic acid, 6-Hydroxypyridine-2-carboxylic acid
M-Clarity™ quality level = MQ100
384305-25G
384305-5G
SIAL384305-25GEA
10060
CZK
SIAL384305-25G
SIAL384305-5G
6-Hydroxypicolinic acid, Sigma-Aldrich®
6-Hydroxypicolinic acid
Vzorec:
C₆H₅NO₃ MW: 139.11 |
MDL Number:
MFCD00192220 CAS číslo: 19621-92-2 EINECS: 417-690-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...