Tisk…

N-Methylisopropylamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-Isopropylmethylamine

M-Clarity™ quality level = MQ200

359378-10ML 359378-50ML 359378-5ML
SIAL359378-10MLEA 2750 CZK
SIAL359378-10ML SIAL359378-50ML SIAL359378-5ML
N-Methylisopropylamine, Sigma-Aldrich®
N-Methylisopropylamine
Vzorec: C₄H₁₁N
MDL Number: MFCD00042859
CAS číslo: 4747-21-1
EINECS: 225-266-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR