Tisk…

Diphenyl chlorophosphate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Diphenyl phosphorochloridate

M-Clarity™ quality level = MQ100

344249-100G 344249-500G
SIAL344249-100GEA 2700 CZK
SIAL344249-100G SIAL344249-500G
Diphenyl chlorophosphate, Sigma-Aldrich®
Diphenyl chlorophosphate
Vzorec: C₁₂H₁₀ClO₃P
MW: 268.64
MDL Number: MFCD00003030
CAS číslo: 2524-64-3
EINECS: 219-759-6
UN: 3265
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR