Tisk…

Cysteamine, Sigma-Aldrich®

Dodavatel: Merck

30070-10G 30070-50G
SIAL30070-10GEA 5650 CZK
SIAL30070-10G SIAL30070-50G
Cysteamine, Sigma-Aldrich®
Cysteamine
Vzorec: C₂H₇NS
MDL Number: MFCD00008196
CAS číslo: 60-23-1
EINECS: 205-858-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR