Dodecyl methacrylate stabilised, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Dodecyl 2-methylacrylate, Lauryl methacrylate
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200
291811-100ML
291811-500ML
SIAL291811-100MLEA
2610
CZK
SIAL291811-100ML
SIAL291811-500ML
Dodecyl methacrylate stabilised, Sigma-Aldrich®
Dodecyl methacrylate
Vzorec:
C₁₆H₃₀O₂ MW: 254.41 |
MDL Number:
MFCD00008972 CAS číslo: 142-90-5 EINECS: 205-570-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...