2-Amino-2-thiazoline hydrochloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
263729-250G
263729-5G
SIAL263729-250GEA
14120
CZK
SIAL263729-250G
SIAL263729-5G
2-Amino-2-thiazoline hydrochloride, Sigma-Aldrich®
2-Amino-2-thiazoline hydrochloride
Vzorec:
C₃H₆N₂S MW: 102,16 g/mol |
MDL Number:
MFCD00012709 CAS číslo: 3882-98-2 EINECS: 217-224-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...