Tisk…

5,6-Dihydro-2H-pyran-2-one, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

262102-10G 262102-1G
SIAL262102-10GEA 17160 CZK
SIAL262102-10G SIAL262102-1G
5,6-Dihydro-2H-pyran-2-one, Sigma-Aldrich®
5,6-Dihydro-2H-pyran-2-one
Vzorec: C₅H₆O₂
MW: 98,1 g/mol
MDL Number: MFCD00010439
CAS číslo: 3393-45-1
UN: 3272
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR