Tisk…
Glyoxylic acid 50% in aqueous solution, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 2-Oxoacetic acid
M-Clarity™ quality level = MQ200
260150-1KG-A 260150-250G-A
SIAL260150-1KG-AEA 3470 CZK
SIAL260150-1KG-A SIAL260150-250G-A
Glyoxylic acid 50% in aqueous solution, Sigma-Aldrich®
Glyoxylic acid
Vzorec: C₂H₂O₃
MW: 74.04 g/mol
MDL Number: MFCD00006958
CAS číslo: 298-12-4
EINECS: 206-058-5
UN: 3265
ADR: 8 III