Tisk…

Hexaethylene glycol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

259268-25G 259268-5G
SIAL259268-25GEA 15710 CZK
SIAL259268-25G SIAL259268-5G
Hexaethylene glycol, Sigma-Aldrich®
Hexaethylene glycol
Vzorec: C₁₂H₂₆O₇
MDL Number: MFCD00002877
CAS číslo: 2615-15-8
EINECS: 220-045-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR