4-Hydrazinobenzoic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Hydrazinylbenzoic acid
246395-25G
246395-5G
SIAL246395-25GEA
4220
CZK
SIAL246395-25G
SIAL246395-5G
4-Hydrazinobenzoic acid, Sigma-Aldrich®
4-Hydrazinobenzoic acid
Vzorec:
C₇H₈N₂O₂ MW: 152,15 g/mol |
MDL Number:
MFCD00007581 CAS číslo: 619-67-0 EINECS: 210-609-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...