Tisk…
Platina 3% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
237558-25G 237558-5G
SIAL237558-25GEA 14330 CZK
SIAL237558-25G SIAL237558-5G
Platina 3% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Platina
Vzorec: Pt
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
EINECS: 231-116-1