Tisk…

Natrium-3-nitrobenzen-1-sulfonát, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sodium 3-nitrobenzenesulphonate, 3-Nitrobenzenesulphonic acid, sodium salt

225193-100G 225193-500G
SIAL225193-100GEA 981 CZK
SIAL225193-100G SIAL225193-500G
Natrium-3-nitrobenzen-1-sulfonát, Sigma-Aldrich®
Natrium-3-nitrobenzen-1-sulfonát
Vzorec: C₆H₄NNaO₅S
MW: 225,16 g/mol
MDL Number: MFCD00007490
CAS číslo: 127-68-4
EINECS: 204-857-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR