Tisk…
Platina 10% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
205958-10G 205958-1G 205958-50G
SIAL205958-10GEA 10570 CZK
SIAL205958-10G SIAL205958-1G SIAL205958-50G
Platina 10% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Platina
Vzorec: Pt
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
EINECS: 231-116-1
UN: 3088
ADR: 4.2,III