Platina 5% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Platinium
MQ100
205931-10G
205931-1G
205931-50G
SIAL205931-10GEA
6990
CZK
SIAL205931-10G
SIAL205931-1G
SIAL205931-50G
Platina 5% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Platina
Vzorec:
Pt MW: 195.08 Bod varu: 3827 Bod tání: 1769 |
MDL Number:
MFCD00011179 CAS číslo: 7440-06-4 EINECS: 231-116-1 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...