Tisk…
Paladium 5% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
205680-100G 205680-10G 205680-50G
SIAL205680-100GEA 30440 CZK
SIAL205680-100G SIAL205680-10G SIAL205680-50G
Paladium 5% on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Palladium
Vzorec: Pd
MW: 106.420 g/mol
MDL Number: MFCD00011167
CAS číslo: 7440-05-3
EINECS: 231-115-6