Cyclohexanecarbonitrile, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
168041-100ML
168041-25ML
SIAL168041-100MLEA
4580
CZK
SIAL168041-100ML
SIAL168041-25ML
Cyclohexanecarbonitrile, Sigma-Aldrich®
Cyclohexanecarbonitrile
Vzorec:
C₇H₁₁N MW: 109,17 g/mol |
MDL Number:
MFCD00040444 CAS číslo: 766-05-2 EINECS: 212-157-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...