Tisk…

Methyl-4-nitrobenzenesulphonate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 4-nitrobenzenesulphonate

160954-10G 160954-1G
SIAL160954-10GEA 11720 CZK
SIAL160954-10G SIAL160954-1G
Methyl-4-nitrobenzenesulphonate, Sigma-Aldrich®
Methyl-4-nitrobenzenesulphonate
Vzorec: C₇H₇NO₅S
MW: 217.2
MDL Number: MFCD00007344
CAS číslo: 6214-20-6
EINECS: 228-282-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR