Tisk…

Acetamidine hydrochloride, Sigma-Aldrich®

Dodavatel: Merck


MQ100

159158-100G 159158-500G
SIAL159158-100GEA 1750 CZK
SIAL159158-100G SIAL159158-500G
Acetamidine hydrochloride, Sigma-Aldrich®
Acetamidine hydrochloride
Vzorec: C₂H₇ClN₂
MW: 94.54
MDL Number: MFCD00013016
CAS číslo: 124-42-5
EINECS: 204-700-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR