Tisk…

Hydantoin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Imidazolidine-2,4-dione

M-Clarity™ quality level = MQ100

156361-100G 156361-500G
SIAL156361-100GEA 1120 CZK
SIAL156361-100G SIAL156361-500G
Hydantoin, Sigma-Aldrich®
Hydantoin
Vzorec: C₃H₄N₂O₂
MW: 100,08 g/mol
MDL Number: MFCD00005259
CAS číslo: 461-72-3
EINECS: 207-313-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR