Tetraethylene glycol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol, Tetraglycol
110175-100G
110175-1KG
110175-20KG
110175-3KG
SIAL110175-100GEA
2530
CZK
SIAL110175-100G
SIAL110175-1KG
SIAL110175-20KG
SIAL110175-3KG
Tetraethylene glycol, Sigma-Aldrich®
Tetraethylene glycol
Vzorec:
C₈H₁₈O₅ MW: 194,23 g/mol |
MDL Number:
MFCD00002879 CAS číslo: 112-60-7 EINECS: 203-989-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...