1,1'-Dimethylferrocene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
109576-25G
109576-5G
SIAL109576-25GEA
7790
CZK
SIAL109576-25G
SIAL109576-5G
1,1'-Dimethylferrocene, Sigma-Aldrich®
1,1'-Dimethylferrocene
Vzorec:
C₁₂H₁₄Fe MW: 214,09 g/mol |
MDL Number:
MFCD00001425 CAS číslo: 1291-47-0 EINECS: 215-067-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...