Chlorid hlinitý, ultra dry ≥99.99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Aluminum chloride
Ampuled under argon
14552.06
14552.14
14552.22
14552.06EA
3000
CZK
14552.06
14552.14
14552.22
Chlorid hlinitý, ultra dry ≥99.99% (metals basis)
Chlorid hlinitý
Purity: 99.99%
CAS No.: 7446-70-0
Molecular Formula: AlCl3
Formula Weight: 133.34
Physical Form: Powder, Ampouled under argon
Density: 2.44
pH: 2,4
Melting Point: 190° subl.
Merck Reference: 14,337
UN No.: 1726
MDL No.: MFCD00003422
Vzorec:
AlCl₃ MW: 133.34 Bod varu: 180 Bod tání: 190 °C (subl.) Hustota: 2.44 Storage Temperature: Ambient |
MDL Number:
MFCD00003422 CAS číslo: 7446-70-0 EINECS: 231-208-1 UN: 1726 ADR: 8 II Merck Index: 14,337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...