Chlorid hlinitý, anhydrous, Reagent Grade
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Aluminum chloride
CAS No.: 7446-70-0
Molecular Formula: AlCl3
Formula Weight: 133.34
Physical Form: Lump/Powder
Density: 2.44
Sensitivity: Moisture Sensitive
Appearance: Yellow to gray
Odour: Pungent
pH: 2,4
Melting Point: 190° subl.
Solubility: Reacts violently with water with liberation of heat; soluble in HCl, ether, ethanol
Merck Reference: 14,337
UN No.: 1726
MDL No.: MFCD00003422
Petroleum refining, as acid catalyst, nucleating agent for titanium dioxide pigment, manufacturing of rubbers and lubricants
Vzorec:
AlCl₃ Bod varu: 180 °C (1013 hPa) Bod tání: 190 °C (subl.) Hustota: 2,44 Storage Temperature: Ambient |
MDL Number:
MFCD00003422 CAS číslo: 7446-70-0 EINECS: 231-208-1 UN: 1726 ADR: 8,II Merck Index: 14,337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...