(R)-(+)-Hydroxypyrrolidine 99%, ee 99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(R)-Pyrrolidin-3-ol, (R)-(+)-3-Pyrrolidinol, (R)-(+)-3-Hydroxypyrrolidine
L19499.03
L19499.06
L19499.03EA
1870
CZK
L19499.03
L19499.06
(R)-(+)-Hydroxypyrrolidine 99%, ee 99%
(R)-(+)-Hydroxypyrrolidine
Purity: 99%
CAS No.: 2799-21-5
Molecular Formula: C4H9NO
Formula Weight: 87.12
Flash Point:
Vzorec:
C₄H₉NO MW: 87,12 g/mol Bod varu: 108…110 °C (8 mmHg) Bod tání: 15 °C Hustota: 1,048 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00145220 CAS číslo: 2799-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...