Tisk…

(S)-(+)-β-Aminophenethyl alcohol ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: (S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol

L13265.03 L13265.06 L13265.14
L13265.03EA 1180 CZK
L13265.03 L13265.06 L13265.14
(S)-(+)-β-Aminophenethyl alcohol ≥98%
(S)-(+)-β-Aminophenethyl alcohol

Purity: 98+% CAS No.: 20989-17-7 Molecular Formula: C8H11NO Formula Weight: 137.18 Melting Point: 76-78° Beilstein Reference: 3196190

Vzorec: C₈H₁₁NO
MW: 137,18 g/mol
Bod tání: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00064404
CAS číslo: 20989-17-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR