Ethylene glycol monoisopropyl ether ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Isopropoxyethanol
L13241.AE
L13241.AU
L13241.0F
L13241.AEEA
903
CZK
L13241.AE
L13241.AU
L13241.0F
Ethylene glycol monoisopropyl ether ≥99%
Ethylene glycol monoisopropyl ether
Purity: 99%
CAS No.: 109-59-1
Molecular Formula: C5H12O2
Formula Weight: 104.15
Flash Point: 45°(113°F)
Refractive Index: 1.4100
Melting Point: -60°
Boiling Point: 143°
Beilstein Reference: 1732184
UN No.: 1992
MDL No.: MFCD00002866
Vzorec:
C₅H₁₂O₂ MW: 104,15 g/mol Bod varu: 143 °C Bod tání: –60 °C Hustota: 0,903 Bod vzplanutí: 45 °C (113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002866 CAS číslo: 109-59-1 EINECS: 203-685-6 UN: 3271 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...