Sodium methanesulphinate 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Methanesulphinic acid, sodium salt
L11360.06
L11360.14
L11360.03
L11360.06EA
2650
CZK
L11360.06
L11360.14
L11360.03
Sodium methanesulphinate 95%
Sodium methanesulphinate
Purity: 95%
CAS No.: 20277-69-4
Molecular Formula: CH3NaO2S
Formula Weight: 102.09
Melting Point: 227-230°
Beilstein Reference: 3565430
Vzorec:
CH₃NaO₂S MW: 102.09 Bod tání: 227…230 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040392 CAS číslo: 20277-69-4 EINECS: 243-669-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...