Tisk…

Acryloyl chloride 96% stabilizovaný

Dodavatel: Thermo Scientific
Nebezpečí


Stabilization: Stabilised with phenothiazine

L10363.18 L10363.09 L10363.30 AB172729.0050
L10363.18EA 2470 CZK
L10363.18 L10363.09 L10363.30 ABCRAB172729.0050
Acryloyl chloride 96% stabilizovaný
Acryloyl chloride

Stabilized with 400 ppm phenothiazine


Purity: 96% CAS No.: 814-68-6 Molecular Formula: C3H3ClO Formula Weight: 90.51 Storage Temperature: -30°C to -10°C Flash Point: -4°(24°F) Refractive Index: 1.4350 Odour: Acrid Boiling Point: 74-76° Beilstein Reference: 635744 UN No.: 3383 MDL No.: MFCD00000731

Vzorec: C₃H₃ClO
MW: 90,51 g/mol
Bod varu: 74…76 °C
Hustota: 1,114
Bod vzplanutí: –4 °C (24 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000731
CAS číslo: 814-68-6
EINECS: 212-399-0
UN: 3383
ADR: 6.1,I

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR