Tisk…

Cyclohexene-1-carbonitrile ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1-Cyclohexene-1-carbonitrile, Cyclohex-1-enecarbonitrile

L10284.09 L10284.04
L10284.09EA 6110 CZK
L10284.09 L10284.04
Cyclohexene-1-carbonitrile ≥98%
Cyclohexene-1-carbonitrile

Purity: 98% CAS No.: 1855-63-6 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 83°(181°F) Refractive Index: 1.4820 Boiling Point: 85-88°/17mm Beilstein Reference: 385837 UN No.: 3276 MDL No.: MFCD00013772

Vzorec: C₇H₉N
MW: 107,16 g/mol
Bod varu: 85…88 °C (17 mmHg)
Hustota: 0,947
Bod vzplanutí: 83 °C (181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013772
CAS číslo: 1855-63-6
EINECS: 217-454-2
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR