Methyl-2-methylnicotinate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
methyl 2-methylpyridine-3-carboxylate, Methyl 2-methylnicotinate
L09074.06
L09074.14
L09074.06EA
2650
CZK
L09074.06
L09074.14
Methyl-2-methylnicotinate ≥97%
Methyl-2-methylnicotinate
Purity: 97%
CAS No.: 65719-09-7
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Refractive Index: 1.5165
Boiling Point: 102-104°/10mm
MDL No.: MFCD00051961
Vzorec:
C₈H₉NO₂ Bod varu: 102…104 °C (10 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00051961 CAS číslo: 65719-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...