3-Hydroxypicolinic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L07489.06
L07489.03
L07489.06EA
3950
CZK
L07489.06
L07489.03
3-Hydroxypicolinic acid ≥98%
3-Hydroxypicolinic acid
Purity: 98%
CAS No.: 874-24-8
Molecular Formula: C6H5NO3
Formula Weight: 139.11
Melting Point: ca 214° dec.
Beilstein Reference: 118954
Vzorec:
C₆H₅NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00006294 CAS číslo: 874-24-8 EINECS: 212-859-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...