N,N-Dimethyl-1,3-diaminopropan 99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L03539.AK
L03539.AU
L03539.AKEA
851
CZK
L03539.AK
L03539.AU
N,N-Dimethyl-1,3-diaminopropan 99%
N,N-Dimethyl-1,3-diaminopropan
Purity: 99%
CAS No.: 109-55-7
Molecular Formula: C5H14N2
Formula Weight: 102.18
Flash Point: 32°(89°F)
Refractive Index: 1.4350
Odour: Ammonia-like
Melting Point: -70°
Boiling Point: 126-128°
Beilstein Reference: 605293
UN No.: 2734
MDL No.: MFCD00008216
Vzorec:
C₅H₁₄N₂ Bod varu: 126…128 °C Bod tání: –70 °C Hustota: 0,818 Bod vzplanutí: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008216 CAS číslo: 109-55-7 EINECS: 203-680-9 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...