Tisk…

5,5-Dimethylhydantoin ≥97%

Dodavatel: Thermo Fisher Scientific

L03388.22 L03388.36
L03388.22EA 865 CZK
L03388.22 L03388.36
5,5-Dimethylhydantoin ≥97%
5,5-Dimethylhydantoin

Purity: 97% CAS No.: 77-71-4 Molecular Formula: C5H8N2O2 Formula Weight: 128.13 Flash Point: 193°(379°F) Odour: Odourless Beilstein Reference: 2827 MDL No.: MFCD00005266

Vzorec: C₅H₈N₂O₂
Bod tání: 174…178 °C
Bod vzplanutí: 193 °C (379 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005266
CAS číslo: 77-71-4
EINECS: 201-051-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR