Dichlorodiethylsilane 94%, rods
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L03239.09
L03239.18
L03239.09EA
1360
CZK
L03239.09
L03239.18
Dichlorodiethylsilane 94%, rods
Dichlorodiethylsilane
Purity: 94%
CAS No.: 1719-53-5
Molecular Formula: C4H10Cl2Si
Formula Weight: 157.12
Flash Point: 28°(82°F)
Refractive Index: 1.4310
Odour: Acrid
Melting Point: -97°
Boiling Point: 129-131°
Beilstein Reference: 605313
UN No.: 1767
MDL No.: MFCD00000493
Vzorec:
C₄H₁₀Cl₂Si Bod varu: 129…131 °C Bod tání: –97 °C Hustota: 1,055 Bod vzplanutí: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000493 CAS číslo: 1719-53-5 EINECS: 217-005-0 UN: 1767 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...