Tisk…

3,4-Dihydro-2H-pyran ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

L02731.AC L02731.AE L02731.AP
L02731.ACEA 913 CZK
L02731.AC L02731.AE L02731.AP
3,4-Dihydro-2H-pyran ≥99%
3,4-Dihydro-2H-pyran

Purity: 99% CAS No.: 110-87-2 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: -15°(5°F) Refractive Index: 1.4420 Odour: Unpleasant pH: 7.0 Melting Point: -70° Boiling Point: 85-86° Beilstein Reference: 103493 UN No.: 2376 MDL No.: MFCD00006558

Vzorec: C₅H₈O
MW: 84,12 g/mol
Bod varu: 85…86 °C
Bod tání: –70 °C
Hustota: 0,926
Bod vzplanutí: –15 °C (5 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006558
CAS číslo: 110-87-2
EINECS: 203-810-4
UN: 2376
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR