Tisk…

Hexaethylene glycol 96%

Dodavatel: Thermo Fisher Scientific

Synonyms: 3,6,9,12,15-Pentaoxaheptadecane-1,17-diol

L02161.06 L02161.14
L02161.06EA 2330 CZK
L02161.06 L02161.14
Hexaethylene glycol 96%
Hexaethylene glycol

Purity: 96% CAS No.: 2615-15-8 Molecular Formula: H(OCH2CH2)6OH Formula Weight: 282.33 Flash Point: 110°(230°F) Refractive Index: 1.4640 Melting Point: 5-7° Boiling Point: 216-217°/4mm Beilstein Reference: 1638281 MDL No.: MFCD00002877

Vzorec: C₁₂H₂₆O₇
MW: 282.33
Bod varu: 216…217 °C (4 mmHg)
Bod tání: 5…7 °C
Hustota: 1.123
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002877
CAS číslo: 2615-15-8
EINECS: 220-045-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR