Tisk…

2-Methyl-2-thiazoline ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Methyl-4,5-dihydrothiazole

L01329.14 L01329.22
L01329.14EA 1800 CZK
L01329.14 L01329.22
2-Methyl-2-thiazoline ≥97%
2-Methyl-2-thiazoline

Purity: 97% CAS No.: 2346-00-1 Molecular Formula: C4H7NS Formula Weight: 101.17 Storage Temperature: 2°C to 8°C Flash Point: 37°(98°F) Refractive Index: 1.5200 Melting Point: -101° Boiling Point: 144-145° Beilstein Reference: 104274 UN No.: 1993 MDL No.: MFCD00005314

Vzorec: C₄H₇NS
MW: 101,17 g/mol
Bod varu: 144…145 °C
Bod tání: –101 °C
Hustota: 1,072
Bod vzplanutí: 37 °C (98 °F)
MDL Number: MFCD00005314
CAS číslo: 2346-00-1
EINECS: 219-071-6
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR