N-tert-Butylbenzylamine 96%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
N-Benzyl-tert-butylamine, N-Benzyl-2-methylpropan-2-amine
L01146.14
L01146.22
L01146.14EA
2040
CZK
L01146.14
L01146.22
N-tert-Butylbenzylamine 96%
N-tert-Butylbenzylamine
Purity: 96%
CAS No.: 3378-72-1
Molecular Formula: C11H17N
Formula Weight: 163.27
Flash Point: 88°(190°F)
Refractive Index: 1.4970
Melting Point: -30° to -25°
Boiling Point: 218-220°
Beilstein Reference: 507464
UN No.: 2735
MDL No.: MFCD00008798
Vzorec:
C₁₁H₁₇N MW: 163,26 g/mol Bod varu: 218…220 °C Bod tání: –30 °C…–25 °C Hustota: 0,899 Bod vzplanutí: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008798 CAS číslo: 3378-72-1 EINECS: 222-179-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...